Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342281.1 | NP_001320134.1 | 1 - 2 | 395 | 65 | |
NM_119602.2 | NP_567966.1 | 1 - 2 | 373 | 65 | ||
GenBank mRNA | AY050864.1 | - | 1 - 2 | 342 | 65 | |
AY117264.1 | - | 1 - 2 | 128 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |