Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124400.3 | NP_199832.1 | 5 - 6 | 2125 | 116 | |
GenBank mRNA | AY035115.1 | - | 5 - 6 | 2091 | 116 | |
AY085553.1 | - | 5 - 6 | 2032 | 116 | ||
AY113860.1 | - | 5 - 6 | 1935 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |