Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174634.1 | NP_777059.1 | 1 - 2 | 137 | 91 | |
GenBank mRNA | X53556.1 | - | 137 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |