Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045880.2 | NP_001039345.1 | 4 - 5 | 425 | 117 | |
GenBank mRNA | BC110146.1 | - | 4 - 5 | 426 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |