Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075484.1 | NP_001068952.1 | 7 - 8 | 916 | 61 | |
GenBank mRNA | BC118256.1 | - | 7 - 8 | 916 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |