Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065174.7 | NP_497575.2 | 4 - 5 | 500 | 90 | |
NM_065175.7 | NP_497576.1 | 4 - 5 | 491 | 90 | ||
NM_065176.6 | NP_497577.2 | 4 - 5 | 491 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |