Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268602.3 | NP_001255531.1 | 1 - 2 | 131 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |