Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129340.5 | NP_001122812.1 | 14 - 15 | 3017 | 96 | |
NM_001276877.3 | NP_001263806.1 | 10 - 11 | 2439 | 96 | ||
NM_001276878.3 | NP_001263807.1 | 10 - 11 | 2439 | 96 | ||
NM_001276879.3 | NP_001263808.1 | 10 - 11 | 2439 | 96 | ||
NM_001307025.4 | NP_001293954.1 | 14 - 15 | 3017 | 96 | ||
NM_070197.6 | NP_502598.2 | 14 - 15 | 3017 | 96 | ||
NM_070198.5 | NP_502599.2 | 14 - 15 | 3011 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |