Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028473.5 | NP_001023644.1 | 6 - 7 | 1169 | 81 | |
NM_072013.8 | NP_504414.4 | 6 - 7 | 1173 | 81 | ||
NM_072014.7 | NP_504415.2 | 6 - 7 | 1182 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |