Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shw-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368471.3 | NP_001355445.1 | 7 - 8 | 1166 | 98 | |
NM_001380790.1 | NP_001368707.1 | 7 - 8 | 1157 | 98 | ||
NM_001380791.1 | NP_001366842.1 | 7 - 8 | 1191 | 98 | ||
GenBank mRNA | DQ185514.1 | - | 6 - 7 | 1103 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |