Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014108587.2 | XP_013964062.1 | 3 - 4 | 447 | 132 | |
XM_022410901.1 | XP_022266609.1 | 3 - 4 | 395 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |