Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496208.2 | XP_003496256.1 | 3 - 4 | 306 | 149 | |
GenBank mRNA | JI871748.1 | - | 2 - 3 | 129 | 149 | |
JP066059.1 | - | 2 - 3 | 127 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |