Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497764.1 | XP_003497812.1 | 1 - 2 | 377 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |