Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509573.2 | XP_003509621.1 | 6 - 7 | 987 | 67 | |
XM_007650190.1 | XP_007648380.1 | 6 - 7 | 1202 | 67 | ||
XM_007650191.1 | XP_007648381.1 | 5 - 6 | 815 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |