Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504668.2 | XP_003504716.1 | 5 - 6 | 907 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |