Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285618.1 | NP_001272547.1 | 4 - 5 | 906 | 87 | |
XM_013974033.2 | XP_013829487.1 | 5 - 6 | 1025 | 87 | ||
XM_013974034.2 | XP_013829488.1 | 5 - 6 | 1035 | 87 | ||
XM_013974035.2 | XP_013829489.1 | 4 - 5 | 876 | 87 | ||
GenBank mRNA | EF102891.1 | - | 4 - 5 | 906 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |