Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039274.1 | XP_017894763.1 | 2 - 3 | 1241 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |