Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039250.1 | XP_017894739.1 | 2 - 3 | 923 | 70 | |
GenBank mRNA | JO535773.2 | - | 2 - 3 | 560 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |