Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173095.1 | NP_001166566.1 | 3 - 4 | 600 | 66 | |
GenBank mRNA | AF018569.1 | - | 3 - 4 | 600 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Cavia porcellus Probe spans exons |