Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140685.5 | NP_648942.1 | 2 - 3 | 453 | 68 | |
NM_168705.2 | NP_730228.1 | 2 - 3 | 460 | 68 | ||
NM_168706.2 | NP_730229.1 | 2 - 3 | 429 | 68 | ||
GenBank mRNA | AF145619.1 | - | 1 - 2 | 386 | 68 | |
EF439708.1 | - | 2 - 3 | 761 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |