Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103520.3 | NP_001096990.1 | 6 - 7 | 1199 | 70 | |
NM_001103521.3 | NP_001096991.1 | 6 - 7 | 994 | 70 | ||
NM_001103522.3 | NP_001096992.1 | 6 - 7 | 1086 | 70 | ||
NM_001258768.1 | NP_001245697.1 | 6 - 7 | 1060 | 70 | ||
NM_001272653.2 | NP_001259582.1 | 7 - 8 | 1250 | 70 | ||
NM_001272654.2 | NP_001259583.1 | 6 - 7 | 1338 | 70 | ||
NM_001298370.1 | NP_001285299.1 | 5 - 6 | 613 | 70 | ||
NM_078614.5 | NP_511169.1 | 7 - 8 | 1141 | 70 | ||
GenBank mRNA | M83787.1 | - | 7 - 8 | 1139 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |