Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GlcAT-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167003.3 | NP_726910.1 | 1 - 2 | 280 | 58 | |
GenBank mRNA | BT044296.1 | - | 1 - 2 | 245 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |