Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB042251.1 | - | 10 - 11 | 1614 | 62 | |
CF995949.1 | - | 1 - 2 | 78 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |