Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nifk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173288.1 | NP_775395.1 | 3 - 4 | 454 | 94 | |
GenBank mRNA | AF506200.1 | - | 3 - 4 | 454 | 94 | |
BC071545.1 | - | 3 - 4 | 448 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |