Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003630.2 | NP_001003630.2 | 4 - 5 | 629 | 66 | |
XM_005165298.4 | XP_005165355.1 | 4 - 5 | 657 | 66 | ||
GenBank mRNA | BC077087.1 | - | 4 - 5 | 633 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |