Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003456.1 | NP_001003456.1 | 3 - 4 | 412 | 78 | |
GenBank mRNA | BC078373.1 | - | 3 - 4 | 412 | 78 | |
BC164925.1 | - | 3 - 4 | 341 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |