Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017758.1 | NP_001017758.1 | 4 - 5 | 530 | 55 | |
GenBank mRNA | BC093357.1 | - | 4 - 5 | 530 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |