Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201114.1 | NP_957408.1 | 2 - 3 | 312 | 67 | |
GenBank mRNA | AY330221.1 | - | 2 - 3 | 292 | 67 | |
BC055237.1 | - | 2 - 3 | 312 | 67 | ||
BC161664.1 | - | 2 - 3 | 280 | 67 | ||
BC171389.1 | - | 2 - 3 | 264 | 67 | ||
BC171393.1 | - | 2 - 3 | 264 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |