Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037838.1 | NP_001032927.1 | 4 - 5 | 561 | 88 | |
XM_015290817.2 | XP_015146303.1 | 5 - 6 | 700 | 88 | ||
GenBank mRNA | DQ272298.1 | - | 4 - 5 | 561 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |