Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000255380.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000448020.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000487470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
ENST00000615928.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018367.hg.1 |
NM_000740.3 |
Refseq |