Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
TC2000009821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168593.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000266068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370069.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
ENST00000370077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009821.hg.1 |
NM_012384.4 |
Refseq |