Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008169.hg.1 |
ENST00000336278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008169.hg.1 |
ENST00000467311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008169.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008169.hg.1 |
TC16000565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008168.hg.1 |
ENST00000336278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008168.hg.1 |
ENST00000467311.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008168.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008168.hg.1 |
TC16000565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000562422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
ENST00000562422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191145.hg.1 |
TC1600011420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000360496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000524887.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000525632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000528525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000562422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
ENST00000562422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011420.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191147.hg.1 |
TC1600011420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
ENST00000336278.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
ENST00000467311.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
NM_006750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191148.hg.1 |
TC1600011420.hg.1 |
Clariom D TC |