Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188528.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188528.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188528.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188528.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188528.hg.1 |
TC1700010997.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700188532.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188532.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188532.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700188532.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700188532.hg.1 |
TC1700010997.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
ENST00000311177.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010997.hg.1 |
NM_024017.4 |
Refseq |