Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136154.1 | NP_001129626.1 | 9 - 10 | 1114 | 199 | |
NM_001136155.1 | NP_001129627.1 | 6 - 7 | 727 | 199 | ||
NM_001243428.1 | NP_001230357.1 | 9 - 10 | 1137 | 199 | ||
NM_001243429.1 | NP_001230358.1 | 6 - 7 | 701 | 127 | ||
NM_001243432.2 | NP_001230361.1 | 9 - 10 | 1114 | 199 | ||
NM_004449.4 | NP_004440.1 | 8 - 9 | 1042 | 127 | ||
NM_182918.3 | NP_891548.1 | 7 - 8 | 945 | 199 | ||
XM_017028288.1 | XP_016883777.1 | 6 - 7 | 827 | 127 | ||
GenBank mRNA | AK297807.1 | - | 9 - 10 | 1114 | 199 | |
AK300395.1 | - | 7 - 8 | 935 | 199 | ||
AK301277.1 | - | 5 - 6 | 471 | 199 | ||
AK303518.1 | - | 6 - 7 | 700 | 199 | ||
AK309469.1 | - | 9 - 10 | 1137 | 199 | ||
AY204741.1 | - | 9 - 10 | 1028 | 199 | ||
BC040168.1 | - | 7 - 8 | 924 | 6052140 | 199 | |
KP100545.1 | - | 6 - 7 | 749 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |