Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SH3 domain binding kinase 1 | SBK1 | SBK | Sense | Chr. 16 - 28258687 - 28323849 on NCBI Build GRCh38 | |
Downstream | nuclear pore complex interacting protein family member B6 | NPIPB6 | NPIPB | 77.9kb | Anti-sense | Chr. 16 - 28342517 - 28364971 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |