Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005491.hg.1 |
ENST00000312422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005491.hg.1 |
NM_001004059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005491.hg.1 |
TC11000434.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007601.hg.1 |
ENST00000312422.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007601.hg.1 |
NM_001004059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153770.hg.1 |
ENST00000312422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153770.hg.1 |
NM_001004059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153770.hg.1 |
ENST00000312422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153770.hg.1 |
NM_001004059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153770.hg.1 |
TC1100007601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007601.hg.1 |
ENST00000312422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007601.hg.1 |
NM_001004059.2 |
Refseq |