Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189479.hg.1 |
TC1700011117.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000300458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000419688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000450727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000502517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
ENST00000576150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011117.hg.1 |
NR_073199.1 |
Refseq |