Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adenylate kinase 5 | AK5 | AK6 | -119.5kb | Anti-sense | Chr. 1 - 77281908 - 77559969 on NCBI Build GRCh38 |
Downstream | ubiquitin specific peptidase 33 | USP33 | VDU1 | 13.3kb | Sense | Chr. 1 - 77695987 - 77759879 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |