Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005552066.2 | XP_005552123.1 | 2 - 3 | 2241 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |