Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_001039169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_001039169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_001039170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_001039170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_023314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
NM_023314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006796.mm.1 |
TC0100000811.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_001039169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_001039169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_001039170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_001039170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_023314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
NM_023314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006804.mm.1 |
TC0100000811.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_001039169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_001039169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_001039170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_001039170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_023314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
NM_023314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006803.mm.1 |
TC0100000811.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006794.mm.1 |
NM_001039170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006794.mm.1 |
NM_001039170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006794.mm.1 |
NM_023314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006794.mm.1 |
NM_023314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006794.mm.1 |
TC0100000811.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_001039169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_001039169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_001039170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_001039170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_023314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
NM_023314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100006798.mm.1 |
TC0100000811.mm.1 |
MTA TC |