Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
737022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
14355 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739593 |
AK013463.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739593 |
AK017306.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
739593 |
XM_001475056.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
349575 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
552036 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857915 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333652 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773068 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198766 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
446384 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291810 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
125142 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708725 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84862 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
65243 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
42172 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P153689 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143753 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310550 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310537 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310532 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310546 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310530 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310541 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310543 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310549 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310533 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310536 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310557 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310553 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310527 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310539 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310544 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310529 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310547 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310535 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310542 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310552 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22310538 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P143753 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04979312 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P153689 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04979302 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00693830 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
BC052072.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
BC082784.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
AF273750.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
AK078394.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P128065 |
NM_033573.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122003 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P153689 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00693830 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P143753 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02891618 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02891621 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02891622 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03185629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310515 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310401 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310368 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310362 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310406 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310500 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310447 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310417 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310359 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310508 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310513 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310549 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22310483 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310558 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310513 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310516 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02891623 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02891620 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02609915 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03043958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310501 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310554 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310495 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02891619 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310518 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310520 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310903 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310560 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310419 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310925 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02609907 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310399 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02609910 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310409 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310526 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310533 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02330633 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310408 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03185626 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310552 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02805027 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310510 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02699705 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03043959 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310539 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310431 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03077468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03076923 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310535 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310906 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02330634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310920 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02618654 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310922 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310546 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310525 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02936730 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03122960 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03185629 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02805028 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310508 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310423 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310550 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02506858 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310415 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310425 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310559 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02976898 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310507 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02310914 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22310511 |
- |
- |