Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020616.1 | NP_065641.1 | 4 - 5 | 460 | 86 | |
GenBank mRNA | AK086219.1 | - | 4 - 5 | 493 | 86 | |
AK159903.1 | - | 4 - 5 | 461 | 86 | ||
AK163012.1 | - | 4 - 5 | 507 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |