Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133770.2 | NP_598531.2 | 11 - 12 | 1204 | 65 | |
GenBank mRNA | AK002619.1 | - | 11 - 12 | 1194 | 65 | |
AK079366.1 | - | 1 - 2 | 248 | 65 | ||
BC013485.1 | - | 8 - 9 | 767 | 4242474 | 65 | |
BC093498.1 | - | 10 - 11 | 1141 | 3669346 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |