Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004884.1 | - | 4 - 5 | 483 | 76 | |
AX775759.1 | - | 5 - 6 | 600 | 76 | ||
BC053099.1 | - | 9 - 10 | 986 | 76 | ||
BC064442.1 | - | 9 - 10 | 954 | 76 | ||
BC082999.1 | - | 2 - 3 | 344 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |