Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282021.1 | NP_001268950.1 | 2 - 3 | 511 | 59 | |
NM_001282022.1 | NP_001268951.1 | 2 - 3 | 511 | 59 | ||
NM_009499.3 | NP_033525.2 | 2 - 3 | 511 | 59 | ||
NR_104069.1 | 2 - 3 | 511 | 59 | |||
GenBank mRNA | AF084548.1 | - | 2 - 3 | 478 | 59 | |
AK140329.1 | - | 2 - 3 | 475 | 59 | ||
AK150103.1 | - | 2 - 3 | 473 | 59 | ||
AK155480.1 | - | 2 - 3 | 473 | 59 | ||
AK157021.1 | - | 2 - 3 | 475 | 59 | ||
AK170780.1 | - | 2 - 3 | 467 | 59 | ||
AK171302.1 | - | 2 - 3 | 475 | 59 | ||
AK171715.1 | - | 2 - 3 | 475 | 59 | ||
BC010223.1 | - | 2 - 3 | 213 | 3500587 | 59 | |
BC015289.1 | - | 2 - 3 | 454 | 4240907 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |