Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013766.2 | NP_038794.1 | 1 - 2 | 74 | 93 | |
GenBank mRNA | AB019118.1 | - | 1 - 2 | 71 | 93 | |
AF150740.1 | - | 1 - 2 | 84 | 93 | ||
AF234637.1 | - | 1 - 2 | 74 | 93 | ||
BC120576.1 | - | 1 - 2 | 62 | 40129499 | 93 | |
BC120578.1 | - | 1 - 2 | 62 | 40129501 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |