Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029738.2 | NP_084014.1 | 4 - 5 | 522 | 73 | |
GenBank mRNA | AK011846.1 | - | 5 - 6 | 550 | 73 | |
AK134648.1 | - | 4 - 5 | 489 | 73 | ||
AK166050.1 | - | 4 - 5 | 603 | 73 | ||
BC024910.1 | - | 4 - 5 | 522 | 3603454 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |