Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2123486 |
NM_001081292.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104562 |
BC078445.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104562 |
BC046514.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104562 |
BC057197.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104562 |
NM_001081292.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
216513 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437913 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
424959 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
146564 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464752 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
250998 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
779351 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
718450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
856797 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
76531 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
608907 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
AK148483.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
BC021452.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
D50494.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
NM_001110826.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
NM_007841.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
808686 |
NM_181324.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
36197 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
78956 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140166 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2123486 |
NM_001081292.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104562 |
BC078445.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104562 |
BC046514.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104562 |
BC057197.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104562 |
NM_001081292.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01489080 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140166 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P301591 |
BC078445.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P301591 |
BC046514.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P301591 |
BC057197.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P301591 |
NM_001081292.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P377667 |
BC078445.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P377667 |
AK053830.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P377667 |
NM_001081292.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140166 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06626839 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06626833 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018365 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018347 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018397 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018410 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018502 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018385 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018345 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018340 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018479 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018381 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018349 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018351 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018394 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018506 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018383 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018404 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018392 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018388 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018361 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018374 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018425 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018363 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018491 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018414 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018355 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018369 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018471 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018460 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018396 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25018429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018491 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018502 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018392 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018407 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018471 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06443400 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018412 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018485 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06593369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018408 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018428 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018425 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018504 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018501 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018487 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018488 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018473 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018499 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018476 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018427 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018497 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05936630 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06457749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05936615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018404 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018414 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018444 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018484 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018450 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018449 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06343250 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018394 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018482 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06626841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05936611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018397 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25018478 |
- |
- |