Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178609.4 | NP_848724.2 | 6 - 7 | 1205 | 112 | |
GenBank mRNA | AK031078.1 | - | 6 - 7 | 1203 | 112 | |
AK041481.1 | - | 6 - 7 | 1203 | 112 | ||
AK045040.1 | - | 6 - 7 | 1170 | 112 | ||
AK143686.1 | - | 6 - 7 | 1178 | 112 | ||
AY463359.1 | - | 6 - 7 | 1202 | 112 | ||
BC145429.1 | - | 6 - 7 | 1158 | 9053987 | 112 | |
BC150772.1 | - | 6 - 7 | 1158 | 9087683 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |