Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010764.2 | NP_034894.2 | 6 - 7 | 971 | 71 | |
GenBank mRNA | AK004817.2 | - | 6 - 7 | 928 | 71 | |
AK035551.1 | - | 6 - 7 | 926 | 71 | ||
AK054315.1 | - | 3 - 4 | 2261 | 71 | ||
AK147928.1 | - | 6 - 7 | 929 | 71 | ||
AK150154.1 | - | 3 - 4 | 303 | 71 | ||
AK170848.1 | - | 3 - 4 | 302 | 71 | ||
AK171166.1 | - | 6 - 7 | 929 | 71 | ||
AK171377.1 | - | 3 - 4 | 302 | 71 | ||
BC005430.1 | - | 6 - 7 | 916 | 3488681 | 71 | |
BC052517.1 | - | 1 - 2 | 870 | 71 | ||
U29947.1 | - | 6 - 7 | 914 | 71 | ||
U87240.1 | - | 6 - 7 | 905 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |